Learn More
Aviva Systems Biology Corp B3galt2 Antibody - C-terminal region (ARP45540_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45540_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3galt2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-207