Learn More
Aviva Systems Biology Corp B3GALT6 Antibody - C-terminal region (ARP50259_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50259_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GALT6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human B3GALT6. Alternate names: beta3GalT6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-60301