Learn More
Aviva Systems Biology Corp B3GLCT Antibody - middle region (ARP49171_P050) | 100uL
SDP
Catalog No. 5001460035
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GALTL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human B3GALTL. Alternate names: B3GTL, B3Glc-T, Gal-T, beta3Glc-T, B3GLCTApplicationWB. Species reactivity: Dog, Human, Mouse, Rat
Catalog No. 50-014-60035 Supplier Aviva Systems Biology Corp Supplier No. ARP49171_P050
This is a rabbit polyclonal antibody against B3GALTL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human B3GALTL. Alternate names: B3GTL, B3Glc-T, Gal-T, beta3Glc-T, B3GLCTApplicationWB. Species reactivity: Dog, Human, Mouse, Rat