Learn More
Aviva Systems Biology Corp B3gat2 Antibody - C-terminal region (ARP50270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3gat2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: GlcAT-S, Glcats, KIAA1963ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-501