Learn More
Aviva Systems Biology Corp B3GAT3 Antibody - N-terminal region (ARP46864_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46864_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GAT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human B3GAT3. Alternate names: GLCATI, GlcAT-IApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-63062