Learn More
Aviva Systems Biology Corp B3GNT4 Antibody - N-terminal region (ARP48944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3GNT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human B3GNT4. Alternate names: B3GN-T4, beta3Gn-T4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-59969