Learn More
ABclonal Technology B3GNT6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A149751000UL
Encompass_Preferred
The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
Catalog No. 50-001-05421