Learn More
Aviva Systems Biology Corp B3gnt7 Antibody - middle region (ARP49028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B3gnt7. It was validated on Western Blot. . Alternate names: C330001H22Rik, beta-3GnT7ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-394