Learn More
Aviva Systems Biology Corp B4GALNT1 Antibody - N-terminal region (ARP45116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B4GALNT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human B4GALNT1. Alternate names: GALGT, GALNACTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58972