Learn More
Aviva Systems Biology Corp B4GALNT3 Antibody - N-terminal region (ARP49109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B4GALNT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human B4GALNT3. Alternate names: B4GalNac-T3, FLJ16224, FLJ40362ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53572