Learn More
Sino Biological B4GALT1 / GGTB2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11220MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human B4GALT1 (rh B4GALT1; Catalog#11220-H07H; NP_001488.2; Gly 44-Ser 398). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7866