Learn More
Sino Biological B4GALT1 / GGTB2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11220RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B4GALT1 / GGTB2 (rh B4GALT1; Catalog#11220-H07H; NP_001488.2; Gly 44-Ser 398). B4GALT1 specific IgG was purified by human B4GALT1 affinity chromatography.
Catalog No. 50-224-8604