Learn More
Aviva Systems Biology Corp B4GALT3 Antibody - N-terminal region (ARP45532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B4GALT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human B4GALT3. Alternate names: beta4Gal-T3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-205