Learn More
Aviva Systems Biology Corp B4galt6 Antibody - C-terminal region (ARP46498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against B4galt6. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA536803, AU022389ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-262