Learn More
Enzo Life Sciences B581 (5mg). CAS: 149759-96-6
SDP
Supplier:  Enzo Life Sciences BMLG2350005
Encompass_Preferred
A potent and selective, cell-permeable farnesyl transferase inhibitor IC50=21 nM, 30-fold selective over geranylgeranyl transferase. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (5mg/ml) or water (2mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0868