Learn More
Medchemexpress LLC B7-1/CD80 Cynomolgu 50ug
SDP
Catalog No. 5000182427
Encompass_Preferred
CD80 is a costimulatory cytokine for cancer immunity that is activated by binding to CD28 or CTLA-4 Tumor immune evasion occurs when CD80 expression is low The increased expression of CD80 induced by TP53 stimulates anti-tumor immune responses B7-1/CD80 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived B7-1/CD80 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82427 Supplier Medchemexpress LLC Supplier No. HYP7276750UG
CD80 is a costimulatory cytokine for cancer immunity that is activated by binding to CD28 or CTLA-4 Tumor immune evasion occurs when CD80 expression is low The increased expression of CD80 induced by TP53 stimulates anti-tumor immune responses B7-1/CD80 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived B7-1/CD80 protein expressed by HEK293 with C-6 His labeled tag