Learn More
Medchemexpress LLC HY-P7321 10ug Medchemexpress, B7-1/CD80, Fc Chimera (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP732110UG
Encompass_Preferred
Medchemexpress, HY-P7321 10ug B7-1/CD80, Fc Chimera (HEK293-expressed), Human CAS: Recombinant Human B7-1/CD80, Fc Chimera (HEK293-expressed) is a polypeptide chain with the C-termimal human IgG1 Fc fragment produced in HEK293 cells. CD80 is a costimulatory molecule known for its role in T-cell activation and also in regulating normal and malignant B cellsactivity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3145