Learn More
Medchemexpress LLC B7-1/CD80 Human Bi 20ug | 20ug
SDP
Catalog No. 5000187585
Encompass_Preferred
The CD80 protein plays a key role in T lymphocyte activation promoting proliferation and cytokine production through CD28 binding In contrast interaction with CTLA-4 inhibits T cell activation B7-1/CD80 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-1/CD80 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87585 Supplier Medchemexpress LLC Supplier No. HYP7806320UG
The CD80 protein plays a key role in T lymphocyte activation promoting proliferation and cytokine production through CD28 binding In contrast interaction with CTLA-4 inhibits T cell activation B7-1/CD80 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-1/CD80 protein expressed by HEK293 with C-Avi C-His labeled tag