Learn More
Medchemexpress LLC B7-1/CD80 Mouse Bi 100ug | 100ug
SDP
Catalog No. 5000187197
Encompass_Preferred
B7-1/CD80 Protein crucial in activating T lymphocytes engages CD28 or CTLA-4 receptors inducing proliferation and cytokine production This interaction regulates immune responses and influences T lymphocyte activation and function Serving as a key checkpoint B7-1/CD80 orchestrates signaling events controlling the immune system s dynamic responses B7-1/CD80 Protein Mouse (Biotinylated HEK293 His) is the recombinant mouse-derived B7-1/CD80 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87197 Supplier Medchemexpress LLC Supplier No. HYP77523100UG
B7-1/CD80 Protein crucial in activating T lymphocytes engages CD28 or CTLA-4 receptors inducing proliferation and cytokine production This interaction regulates immune responses and influences T lymphocyte activation and function Serving as a key checkpoint B7-1/CD80 orchestrates signaling events controlling the immune system s dynamic responses B7-1/CD80 Protein Mouse (Biotinylated HEK293 His) is the recombinant mouse-derived B7-1/CD80 protein expressed by HEK293 with C-His labeled tag