Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-1 / CD80 Protein, Fc,Avitag™
Supplier:  ACROBiosystems B71H82F2200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human B7-1, Fc,Avitag (B71-H82F2) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # NP_005182.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.3 kDa. As a result of glycosylation, the protein migrates as 65-95 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) with a linear range of 0.6-9.8 ng/mL (QC tested).;Immobilized Human PD-L1, Fc Tag (Cat. No. PD1-H5258) at 10 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) with a linear range of 0.156-5 μg/mL (Routinely tested).
Catalog No. 50-195-7425