Learn More
ACROBiosystems ACROBiosystems Mouse B7-1 / CD80 Protein, His Tag
Catalog No. 501957433
Encompass_Preferred
Recombinant Protein;100UG;Mouse B7-1, His Tag (CD0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 38 - Lys 245 (Accession # Q00609-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-1, His Tag (Cat. No. CD0-M5228) at 2 μg/mL (100 μL/well) can bind Mouse CTLA-4, Fc Tag (Cat. No. CT4-M5256) with a linear range of 0.12-0.5 ng/mL (QC tested).
Catalog No. 50-195-7433 Supplier ACROBiosystems Supplier No. CD0M5228100UG
Recombinant Protein;100UG;Mouse B7-1, His Tag (CD0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 38 - Lys 245 (Accession # Q00609-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-1, His Tag (Cat. No. CD0-M5228) at 2 μg/mL (100 μL/well) can bind Mouse CTLA-4, Fc Tag (Cat. No. CT4-M5256) with a linear range of 0.12-0.5 ng/mL (QC tested).