Learn More
Medchemexpress LLC B7-2/CD86 Human Bi 50ug | 50ug
SDP
Catalog No. 5000187815
Encompass_Preferred
B7-2/CD86 protein negatively regulates T-cell activation by disrupting CD86 cluster formation modulating the T-cell response and influencing immune activation B7-2/CD86 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-2/CD86 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87815 Supplier Medchemexpress LLC Supplier No. HYP7806450UG
B7-2/CD86 protein negatively regulates T-cell activation by disrupting CD86 cluster formation modulating the T-cell response and influencing immune activation B7-2/CD86 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-2/CD86 protein expressed by HEK293 with C-Avi C-His labeled tag