Learn More
Medchemexpress LLC B7-2 (CD86) protein, human, biotinylated | >95.0% | 100 UG
SDP
Catalog No. 5000291401
Encompass_Preferred
Biotinylated human B7-2 (CD86) recombinant protein expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized and intended for use in binding assays, immunoassays, and other applications requiring a biotin-tagged extracellular human protein.
  • Biotinylated for streptavidin-based detection and capture.
  • Expressed in HEK293 cells to preserve human-like post-translational modifications.
  • High purity suitable for sensitive biochemical and immunological assays.
  • Lyophilized formulation with stabilizers for ambient shipping and long-term storage.
  • Reconstitution guidance provided; aliquot and store at -20°C or -80°C for extended stability.
  • Suitable for binding studies, ELISA, and other assay formats requiring a tagged ligand.
Catalog No. 50-002-91401 Supplier Medchemexpress LLC Supplier No. HYP75489100UG
Biotinylated human B7-2 (CD86) recombinant protein expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized and intended for use in binding assays, immunoassays, and other applications requiring a biotin-tagged extracellular human protein.
  • Biotinylated for streptavidin-based detection and capture.
  • Expressed in HEK293 cells to preserve human-like post-translational modifications.
  • High purity suitable for sensitive biochemical and immunological assays.
  • Lyophilized formulation with stabilizers for ambient shipping and long-term storage.
  • Reconstitution guidance provided; aliquot and store at -20°C or -80°C for extended stability.
  • Suitable for binding studies, ELISA, and other assay formats requiring a tagged ligand.