Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, His Tag (HPLC-verified)
Supplier:  ACROBiosystems CD6C52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.3 kDa. The protein migrates as 50-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, Fc Tag (Cat. No. CT4-C5256) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-2, His Tag (HPLC-verified) (Cat. No. CD6-C52H5) with a linear range of 0.312-5 μg/mL (QC tested).;Immobilized Cynomolgus / Rhesus macaque B7-2, His Tag (HPLC-verified) (Cat. No. CD6-C52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CTLA-4, Fc,Avitag (Cat. No. CT4-H82F3) with a linear range of 0.02-0.625 μg/mL (Routinely tested).
Catalog No. 50-195-7531