Learn More
ACROBiosystems ACROBiosystems Human B7-2 / CD86 Protein, His Tag
Supplier:  ACROBiosystems CD6H52231MG
Encompass_Preferred
Recombinant Protein;1MG;Human B7-2, His Tag (CD6-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession # AAH40261).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 42-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) at 5 μg/mL (100 μL/well) can bind Human B7-2, His Tag (Cat. No. CD6-H5223) with a linear range of 0.01-0.156 μg/mL (QC tested).
Catalog No. 50-195-7517