Learn More
MSC Value Collection Stud, B7, 3/4-10 x 4 in OAL, 25/Pk.
Catalog No. 502866245
Encompass
This fully threaded stud is manufactured from steel and engineered for demanding applications. Conforming to ASTM A193 standards, it provides reliable fastening.
  • Made from steel
  • Grade B7 material
  • Thread size: 3/4-10
  • Overall length: 4 in
  • Uncoated finish
  • Thread fit class: 7B
  • Supplied in a package of 25
Catalog No. 50-286-6245 Supplier MSC Supplier No. 80950611
This fully threaded stud is manufactured from steel and engineered for demanding applications. Conforming to ASTM A193 standards, it provides reliable fastening.
  • Made from steel
  • Grade B7 material
  • Thread size: 3/4-10
  • Overall length: 4 in
  • Uncoated finish
  • Thread fit class: 7B
  • Supplied in a package of 25