Learn More
ACROBiosystems ACROBiosystems Human B7-H2 / ICOSLG Protein, Fc Tag
Supplier:  ACROBiosystems B72H5254100UG
Encompass_Preferred
Recombinant Protein;100UG;Human B7-H2, Fc Tag (B72-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession # O75144-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.1 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ICOS (C136S, C137S), His Tag (Cat. No. ICS-H52H6) at 5 μg/mL (100 μL/well) can bind Human B7-H2, Fc Tag (Cat. No. B72-H5254) with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-195-7111