Learn More
ACROBiosystems ACROBiosystems Mouse B7-H2 / ICOSLG Protein, His Tag
Catalog No. 501956732
Encompass_Preferred
Recombinant Protein;1MG;Mouse B7-H2, His Tag (B72-M52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 47 - Lys 279 (Accession # Q9JHJ8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-H2, His Tag (Cat. No. B72-M52H7) at 1 μg/mL (100 μL/well) can bind Mouse ICOS (C137S, C138S), Fc Tag (Cat. No. ICS-M5259) with a linear range of 1-78 ng/mL (QC tested).
Catalog No. 50-195-6732 Supplier ACROBiosystems Supplier No. B72M52H71MG
Recombinant Protein;1MG;Mouse B7-H2, His Tag (B72-M52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 47 - Lys 279 (Accession # Q9JHJ8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-H2, His Tag (Cat. No. B72-M52H7) at 1 μg/mL (100 μL/well) can bind Mouse ICOS (C137S, C138S), Fc Tag (Cat. No. ICS-M5259) with a linear range of 1-78 ng/mL (QC tested).