Learn More
ACROBiosystems ACROBiosystems Human B7-H2 / ICOSLG Protein, His Tag
Catalog No. 501957516
Encompass_Preferred
Recombinant Protein;1MG;Human B7-H2, His Tag (B72-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession # NP_056074).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.8 kDa. The protein migrates as 45-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ICOS, Fc Tag (Cat. No. ICS-H5258) at 1 μg/mL (100 μL/well) can bind Human B7-H2, His Tag (Cat. No. B72-H5221) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-7516 Supplier ACROBiosystems Supplier No. B72H52211MG
Recombinant Protein;1MG;Human B7-H2, His Tag (B72-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession # NP_056074).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.8 kDa. The protein migrates as 45-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ICOS, Fc Tag (Cat. No. ICS-H5258) at 1 μg/mL (100 μL/well) can bind Human B7-H2, His Tag (Cat. No. B72-H5221) with a linear range of 5-78 ng/mL (QC tested).