Learn More
ACROBiosystems ACROBiosystems Cynomolgus B7-H2 / ICOSLG Protein, His Tag
Catalog No. 501956576
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession # XP_005548617.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.5 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ICOS, Fc Tag (Cat. No. ICS-H5258) at 1 μg/mL (100 μL/well) can bind Cynomolgus B7-H2, His Tag (Cat. No. B72-C52Hb) with a linear range of 0.6-20 ng/mL (QC tested).
Catalog No. 50-195-6576 Supplier ACROBiosystems Supplier No. B72C52HB1MG
Recombinant Protein;1MG;Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession # XP_005548617.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.5 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ICOS, Fc Tag (Cat. No. ICS-H5258) at 1 μg/mL (100 μL/well) can bind Cynomolgus B7-H2, His Tag (Cat. No. B72-C52Hb) with a linear range of 0.6-20 ng/mL (QC tested).