Learn More
Medchemexpress LLC B7-H2/ICOSLG Mouse 10ug
SDP
Catalog No. 5000181100
Encompass_Preferred
B7-H2/ICOSLG is an important ligand of the T cell receptor ICOS and mediates immune responses It acts as a costimulatory signal promoting T and B cell proliferation cytokine secretion and plasma cell differentiation B7-H2/ICOSLG Protein Mouse (HEK293 His) is the recombinant mouse-derived B7-H2/ICOSLG protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81100 Supplier Medchemexpress LLC Supplier No. HYP7312010UG
B7-H2/ICOSLG is an important ligand of the T cell receptor ICOS and mediates immune responses It acts as a costimulatory signal promoting T and B cell proliferation cytokine secretion and plasma cell differentiation B7-H2/ICOSLG Protein Mouse (HEK293 His) is the recombinant mouse-derived B7-H2/ICOSLG protein expressed by HEK293 with C-His labeled tag