Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC B7-H2/ICOSLG protein, cynomolgus | >95.0% | 20 UG
SDP

Catalog No. 5000290031
Encompass_Preferred

B7-H2/ICOSLG protein (cynomolgus) is a recombinant extracellular domain of the ICOS ligand produced in HEK293 mammalian cells with a C-terminal His tag. Supplied lyophilized from PBS (pH 7.4), the protein is intended for immunology and receptor-ligand interaction studies and is characterized by low endotoxin and a defined amino acid range (M1-T256).

  • Recombinant protein expressed in HEK293 mammalian cells.
  • C-terminal His tag for purification and detection.
  • Amino acid range M1-T256 corresponding to the extracellular domain.
  • Observed molecular weight 50-70 kDa.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized formulation from PBS, pH 7.4; recommend aliquoting for storage.

Catalog No. 50-002-90031 Supplier Medchemexpress LLC Supplier No. HYP7639320UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

B7-H2/ICOSLG protein (cynomolgus) is a recombinant extracellular domain of the ICOS ligand produced in HEK293 mammalian cells with a C-terminal His tag. Supplied lyophilized from PBS (pH 7.4), the protein is intended for immunology and receptor-ligand interaction studies and is characterized by low endotoxin and a defined amino acid range (M1-T256).

  • Recombinant protein expressed in HEK293 mammalian cells.
  • C-terminal His tag for purification and detection.
  • Amino acid range M1-T256 corresponding to the extracellular domain.
  • Observed molecular weight 50-70 kDa.
  • Endotoxin <1 EU/μg (LAL method).
  • Lyophilized formulation from PBS, pH 7.4; recommend aliquoting for storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.