Learn More
Genscript Corporation B7-H3 4Ig /B7-H3b hFc Chimer
Catalog No. 5000056710
Encompass
B7-H3 a member of the B7 family of immunomodulatory molecules is overexpressed in a wide range of solid cancers B7-H3 binds to activated T cells via an as yet unidentified receptor In assays using sub-optimal amount so anti-CD3 stimulation 2Ig B7 H3 enhances T cell proliferation T cell interferon-gamma (IFN-gamma) production and cytotoxic T cells induction
Catalog No. 50-000-56710 Supplier Genscript Corporation Supplier No. Z03889100
B7-H3 a member of the B7 family of immunomodulatory molecules is overexpressed in a wide range of solid cancers B7-H3 binds to activated T cells via an as yet unidentified receptor In assays using sub-optimal amount so anti-CD3 stimulation 2Ig B7 H3 enhances T cell proliferation T cell interferon-gamma (IFN-gamma) production and cytotoxic T cells induction