Learn More
ACROBiosystems ACROBiosystems Cynomolgus B7-H3 / CD276 Protein, His Tag
Supplier:  ACROBiosystems B73C52HA100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus B7-H3, His Tag (B73-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Glu 465 (Accession # XP_015308534.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus B7-H3, His Tag (Cat. No. B73-C52Ha) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-195-6592