Learn More
ACROBiosystems ACROBiosystems Human B7-H3 (4Ig) / B7-H3b Protein, His Tag
Catalog No. 501957135
Encompass_Preferred
Recombinant Protein;1MG;Human B7-H3 (4Ig), His Tag (B7B-H52E7) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) with a linear range of 1-10 ng/mL (Routinely tested).;Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) inhibits Anti-CD3-induced proliferation of PBMC. The EC50 for this effect is 7.95 μg/mL (Routinely tested).
Catalog No. 50-195-7135 Supplier ACROBiosystems Supplier No. B7BH52E71MG
Recombinant Protein;1MG;Human B7-H3 (4Ig), His Tag (B7B-H52E7) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) with a linear range of 1-10 ng/mL (Routinely tested).;Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) inhibits Anti-CD3-induced proliferation of PBMC. The EC50 for this effect is 7.95 μg/mL (Routinely tested).