Learn More
Medchemexpress LLC HY-P7326 10ug Medchemexpress, B7-H3/ICOSLG, Fc Chimera (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP732610UG
Encompass_Preferred
Medchemexpress, HY-P7326 10ug B7-H3/ICOSLG, Fc Chimera (HEK293-expressed), Human CAS: Recombinant Human B7-H3/ICOSLG, Fc Chimera (HEK293-expressed) is a polypeptide chain containing the C-termimal His tag produced in HEK293 cells. B7-H3 is an immune checkpoint from the B7 family of molecules. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3147