Learn More
Enzo Life Sciences B7-H3 monoclonal antibody (2A7) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7010100
Encompass_Preferred
Clone: 2A7. Isotype: IgG1κ. Immunogen: Extracellular domain of human B7-H3. Species reactivity: Human. Applications: Flow Cytometry, ICC, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3240