Learn More
Sino Biological Recombinant Human B7-H3 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 11188H08HB20
Encompass_Preferred
A DNA sequence encoding the human CD276 (Q5ZPR3-1) extracellular domain (Met1-Thr461) was expressed, with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6053