Learn More
ACROBiosystems ACROBiosystems Human B7-H4 Protein, Fc Tag, low endotoxin
Catalog No. 501957159
Encompass_Preferred
Recombinant Protein;100UG;Human B7-H4, Fc Tag, low endotoxin (B74-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # Q7Z7D3-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 66-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H4, Fc Tag, low endotoxin (Cat. No. B74-H5256) at 0.5 μg/mL (100 μL/well) can bind Anti-B7-H4 MAb (Human IgG1) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-195-7159 Supplier ACROBiosystems Supplier No. B74H5256100UG
Recombinant Protein;100UG;Human B7-H4, Fc Tag, low endotoxin (B74-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # Q7Z7D3-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 66-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H4, Fc Tag, low endotoxin (Cat. No. B74-H5256) at 0.5 μg/mL (100 μL/well) can bind Anti-B7-H4 MAb (Human IgG1) with a linear range of 0.2-8 ng/mL (QC tested).