Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-H4 Protein, Avitag™,His Tag (recommended for biopanning)
Catalog No. 501957426
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (B74-H82E2) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # NP_078902).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 28 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human B7-H4 MAb, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (Cat. No. B74-H82E2) with a linear range of 3-48 ng/mL (QC tested).
Catalog No. 50-195-7426 Supplier ACROBiosystems Supplier No. B74H82E2200UG
Recombinant Protein;200UG;Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (B74-H82E2) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # NP_078902).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 28 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human B7-H4 MAb, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (Cat. No. B74-H82E2) with a linear range of 3-48 ng/mL (QC tested).