Learn More
Medchemexpress LLC B7-H4 Human HEK293 100ug
SDP
Catalog No. 5000178920
Encompass_Preferred
The B7-H4 protein acts as a negative regulator inhibiting T cell-mediated immune responses including activation proliferation cytokine production and cytotoxicity When expressed on tumor macrophages it cooperates with regulatory T cells to suppress tumor-associated antigen-specific T cell immunity B7-H4 Protein Human (HEK293 His) is the recombinant human-derived B7-H4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78920 Supplier Medchemexpress LLC Supplier No. HYP74395100UG
The B7-H4 protein acts as a negative regulator inhibiting T cell-mediated immune responses including activation proliferation cytokine production and cytotoxicity When expressed on tumor macrophages it cooperates with regulatory T cells to suppress tumor-associated antigen-specific T cell immunity B7-H4 Protein Human (HEK293 His) is the recombinant human-derived B7-H4 protein expressed by HEK293 with C-His labeled tag