Learn More
Medchemexpress LLC B7-H4 Mouse HEK293 100ug
SDP
Catalog No. 5000178431
Encompass_Preferred
The B7-H4 protein acts as a negative regulator and inhibits T cell-mediated immune responses including activation proliferation cytokine production and cytotoxicity Significant significance can be observed when expressed on tumor macrophages cooperating with regulatory T cells (Tregs) to suppress tumor-associated antigen-specific T cell immunity B7-H4 Protein Mouse (HEK293 His) is the recombinant mouse-derived B7-H4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78431 Supplier Medchemexpress LLC Supplier No. HYP74394100UG
The B7-H4 protein acts as a negative regulator and inhibits T cell-mediated immune responses including activation proliferation cytokine production and cytotoxicity Significant significance can be observed when expressed on tumor macrophages cooperating with regulatory T cells (Tregs) to suppress tumor-associated antigen-specific T cell immunity B7-H4 Protein Mouse (HEK293 His) is the recombinant mouse-derived B7-H4 protein expressed by HEK293 with C-His labeled tag