Learn More
Genscript Corporation B7-H5/Gi24/VISTABiotin His
Catalog No. 5000059168
Encompass
B7-H5 also known as VISTA B7H5 Dies1 SISP1 and C10orf54 is a 55-65 kDa member of the Ig superfamily It is a transmembrane molecule expressed in bone on embryonic stem cells (ESCs) and on tumor cell surfaces
Catalog No. 50-000-59168 Supplier Genscript Corporation Supplier No. Z04410500
B7-H5 also known as VISTA B7H5 Dies1 SISP1 and C10orf54 is a 55-65 kDa member of the Ig superfamily It is a transmembrane molecule expressed in bone on embryonic stem cells (ESCs) and on tumor cell surfaces