Learn More
Medchemexpress LLC B7-H6 Human Biotin 50ug | 50ug
SDP
Catalog No. 5000187522
Encompass_Preferred
B7-H6 a distinctive monomeric protein uniquely triggers NCR3-dependent natural killer (NK) cell activation specifically interacting with NCR3 while avoiding engagement with other NK cell-activating receptors (NCR1 NCR2 and KLRK1) This specificity underscores B7-H6 s unique role in initiating NK cell responses through the NCR3 pathway within the intricate network of NK cell activation mechanisms B7-H6 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-H6 protein expressed by HEK293 with C-His C-Avi labeled tag
Catalog No. 50-001-87522 Supplier Medchemexpress LLC Supplier No. HYP7807650UG
B7-H6 a distinctive monomeric protein uniquely triggers NCR3-dependent natural killer (NK) cell activation specifically interacting with NCR3 while avoiding engagement with other NK cell-activating receptors (NCR1 NCR2 and KLRK1) This specificity underscores B7-H6 s unique role in initiating NK cell responses through the NCR3 pathway within the intricate network of NK cell activation mechanisms B7-H6 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived B7-H6 protein expressed by HEK293 with C-His C-Avi labeled tag