Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC B7-H6 protein, cynomolgus | >95.0% | 500 UG
SDP

Catalog No. 5000293753
Encompass_Preferred

Recombinant cynomolgus B7-H6 extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag. Provided as a purified protein for use in biochemical and immunological assays involving NCR3/NKp30 interactions, receptor-ligand binding, and cell-based functional studies.

  • Recombinant extracellular domain (aa D25-D259)
  • Expressed in HEK293 mammalian cells
  • C-terminal 6xHis tag for purification and detection
  • Molecular weight ~41-52 kDa due to glycosylation
  • Purity >95% by reducing SDS-PAGE
  • Supplied as 500 μg; certificate of analysis available
  • Suitable for binding and functional assays targeting NCR3/NKp30
  • Recommended to freeze aliquots at -20°C or -80°C for long-term storage

Catalog No. 50-002-93753 Supplier Medchemexpress LLC Supplier No. HYP76743500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cynomolgus B7-H6 extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag. Provided as a purified protein for use in biochemical and immunological assays involving NCR3/NKp30 interactions, receptor-ligand binding, and cell-based functional studies.

  • Recombinant extracellular domain (aa D25-D259)
  • Expressed in HEK293 mammalian cells
  • C-terminal 6xHis tag for purification and detection
  • Molecular weight ~41-52 kDa due to glycosylation
  • Purity >95% by reducing SDS-PAGE
  • Supplied as 500 μg; certificate of analysis available
  • Suitable for binding and functional assays targeting NCR3/NKp30
  • Recommended to freeze aliquots at -20°C or -80°C for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.