Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC B7-H6 protein, cynomolgus (HEK293, His) | 97.0% | 5 UG
SDP

Catalog No. 5000293736
Encompass_Preferred

Recombinant B7-H6 protein from cynomolgus macaque represents the extracellular region (D25-D259) and is produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder for in vitro research, it is intended for binding assays, antibody development, and cell-based studies involving NK cell activation.

  • Recombinant extracellular domain of B7-H6 (residues D25-D259).
  • Produced in HEK293 cells to retain human-like glycosylation.
  • C-terminal His tag for purification and detection.
  • Lyophilized formulation for stable storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight approximately 41-52 kDa due to glycosylation.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Stable storage at -20°C; aliquot and add carrier protein for long-term storage.

Catalog No. 50-002-93736 Supplier Medchemexpress LLC Supplier No. HYP767435UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant B7-H6 protein from cynomolgus macaque represents the extracellular region (D25-D259) and is produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder for in vitro research, it is intended for binding assays, antibody development, and cell-based studies involving NK cell activation.

  • Recombinant extracellular domain of B7-H6 (residues D25-D259).
  • Produced in HEK293 cells to retain human-like glycosylation.
  • C-terminal His tag for purification and detection.
  • Lyophilized formulation for stable storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight approximately 41-52 kDa due to glycosylation.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Stable storage at -20°C; aliquot and add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.