Learn More
ACROBiosystems ACROBiosystems Human B7-H7 / HHLA2 Protein, Fc Tag
Catalog No. 501957056
Encompass_Preferred
Recombinant Protein;1MG;Human B7-H7, Fc Tag (B77-H5257) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.4 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD28H, His Tag (Cat. No. CDH-H52H3) at 5 μg/mL (100 μL/well) can bind Human B7-H7, Fc Tag (Cat. No. B77-H5257) with a linear range of 1-78 ng/mL (QC tested).
Catalog No. 50-195-7056 Supplier ACROBiosystems Supplier No. B77H52571MG
Recombinant Protein;1MG;Human B7-H7, Fc Tag (B77-H5257) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.4 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD28H, His Tag (Cat. No. CDH-H52H3) at 5 μg/mL (100 μL/well) can bind Human B7-H7, Fc Tag (Cat. No. B77-H5257) with a linear range of 1-78 ng/mL (QC tested).