Learn More
Cayman Chemical B71CD80 LNg Isoform EXrac
SDP
Catalog No. 502949340
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 51.7 kDa
Catalog No. 50-294-9340 Supplier Cayman Chemical Supplier No. 31821100 UG
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 51.7 kDa