Learn More
Cayman Chemical B71CD80 LNg Isoform EXrac
SDP
Catalog No. 502949755
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 25.4 kDa
Catalog No. 50-294-9755 Supplier Cayman Chemical Supplier No. 32013100 UG
Source: Recombinant human C-terminal His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 25.4 kDa