Learn More
Cayman Chemical B71CD80 LNg Isoform Extrac 1mg
SDP
Catalog No. 502949397
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 51.7 kDa
Catalog No. 50-294-9397 Supplier Cayman Chemical Supplier No. 318211 MG
Source: Active recombinant C-terminal human IgG1 Fc-His-tagged CD80 expressed in HEK293 cells • Amino acids: 35-242 • MW: 51.7 kDa